Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019454.3 | NP_062327.2 | 7 | 1286 | ||
GenBank mRNA | AB043893.1 | - | 7 | 929 | ||
AF253469.1 | - | 7 | 1241 | |||
AF273454.1 | - | 7 | 1307 | |||
AK004739.1 | - | 7 | 1254 | |||
AK031699.1 | - | 7 | 1255 | |||
AK052322.1 | - | 7 | 1287 | |||
AK165517.1 | - | 7 | 1254 | |||
AK171717.1 | - | 7 | 1253 | |||
BC042497.1 | - | 7 | 1241 | 4017786 | ||
BC049130.1 | - | 7 | 1246 | 6825525 |