Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018801.3 | NP_061271.1 | 8 | 1339 | ||
NM_173067.3 | NP_775090.1 | 9 | 1471 | |||
NM_173068.2 | NP_775091.2 | 10 | 1831 | |||
GenBank mRNA | AB026804.1 | - | 8 | 946 | ||
AB075900.1 | - | 9 | 1078 | |||
AB075901.1 | - | 11 | 1291 | |||
AK075696.1 | - | 7 | 1139 | |||
BC105660.1 | - | 8 | 945 | |||
BC139806.1 | - | 8 | 946 | 40046580 |