Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018811.6 | NP_061281.3 | 9, 10 | 1183 | ||
GenBank mRNA | AF201730.1 | - | 9, 10 | 1172 | ||
AF546701.1 | - | 9, 10 | 1184 | |||
AK008690.1 | - | 9, 10 | 1193 | |||
AK016296.1 | - | 7, 8 | 844 | |||
AK139526.1 | - | 9, 10 | 1192 | |||
BC027098.1 | - | 9, 10 | 1153 | 5356667 | ||
BC089477.1 | - | 9, 10 | 1163 | 6741521 |