Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290624.1 | NP_001277553.1 | 9 | 1708 | ||
GenBank mRNA | AK037217.1 | - | 9 | 1671 | ||
AK083704.1 | - | 8 | 1675 | |||
BC023671.1 | - | 9 | 1704 | |||
BC032173.1 | - | 6 | 2029 | |||
BC049258.1 | - | 9 | 1650 | 5250488 |