Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002274.3 | NP_002265.2 | 5 | 982 | ||
NM_153490.2 | NP_705694.2 | 5 | 982 | |||
GenBank mRNA | AK092276.1 | - | 6 | 947 | ||
AK223051.1 | - | 5 | 981 | |||
AK223077.1 | - | 5 | 981 | |||
AK290244.1 | - | 5 | 981 | |||
AK296806.1 | - | 6 | 770 | |||
AK309752.1 | - | 5 | 983 | |||
BC002661.2 | - | 5 | 966 | 3610361 | ||
BC077718.1 | - | 5 | 959 | 5479174 | ||
BC126184.1 | - | 5 | 946 | 8991900 | ||
BC136494.1 | - | 5 | 946 | 9020483 | ||
DR422220.1 | - | 5 | 465 | |||
X14640.1 | - | 5 | 961 | |||
X52426.1 | - | 5 | 966 |