Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173036.3 | NP_766624.3 | 5, 6 | 701 | ||
GenBank mRNA | AK037149.1 | - | 1, 2 | 1646 | ||
AK154648.1 | - | 5, 6 | 677 | |||
AK155164.1 | - | 5, 6 | 673 | |||
BC026426.1 | - | 3, 4 | 407 | |||
BC064726.1 | - | 5, 6 | 661 | 30433392 |
Set Membership: |
G-protein coupled receptor |