Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012021.2 | NP_036151.1 | 5, 6 | 563 | ||
GenBank mRNA | AF110733.1 | - | 5, 6 | 489 | ||
AF124994.1 | - | 5, 6 | 652 | |||
AF197951.1 | - | 5, 6 | 563 | |||
AK002383.1 | - | 5, 6 | 529 | |||
AK003332.2 | - | 5, 6 | 529 | |||
AK009858.1 | - | 5, 6 | 998 | |||
AK136130.1 | - | 5, 6 | 504 | |||
AK152653.1 | - | 5, 6 | 529 | |||
BC008174.1 | - | 5, 6 | 948 | 3481713 | ||
BC014682.1 | - | 3, 4 | 173 |