Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dfna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018769.3 | NP_061239.1 | 4 | 487 | ||
GenBank mRNA | AF073309.1 | - | 4 | 488 | ||
AK016561.1 | - | 855 | ||||
AK089459.1 | - | 474 | ||||
AK155135.1 | - | 466 | ||||
AK171278.1 | - | 4 | 486 | |||
BC051962.1 | - | 4 | 547 | |||
BC086618.1 | - | 4 | 547 | |||
BC132303.1 | - | 3 | 458 | 40130580 |