Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sertad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018820.4 | NP_061290.1 | 2 | 494 | ||
GenBank mRNA | AF218291.1 | - | 2 | 401 | ||
AF366400.1 | - | 2 | 398 | |||
AF366401.1 | - | 2 | 537 | |||
AK002324.1 | - | 2 | 495 | |||
AK004022.1 | - | 2 | 494 | |||
AK008303.1 | - | 2 | 495 | |||
BC016077.1 | - | 2 | 454 | 4486318 |