Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018864.5 | NP_061352.2 | 9 | 884 | ||
GenBank mRNA | AF042730.1 | - | 9 | 829 | ||
AK011770.1 | - | 9 | 822 | |||
AK078406.1 | - | 9 | 833 | |||
AK083238.1 | - | 9 | 818 | |||
AK145472.1 | - | 821 | ||||
AK147778.1 | - | 9 | 844 | |||
AK152774.1 | - | 9 | 835 | |||
AK153457.1 | - | 9 | 851 | |||
AK160203.1 | - | 9 | 822 | |||
AK160467.1 | - | 9 | 816 | |||
AK165977.1 | - | 9 | 826 | |||
AK168457.1 | - | 9 | 859 | |||
BC049080.1 | - | 8 | 1626 | 6419835 | ||
BC055722.1 | - | 8 | 779 | 6413389 |