Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139520.1 | NP_001132992.1 | 10 | 1273 | ||
NM_018851.3 | NP_061339.3 | 10 | 1273 | |||
GenBank mRNA | AK054490.1 | - | 10 | 1172 | ||
AK078936.1 | - | 10 | 1216 | |||
AK128954.1 | - | 9 | 1071 | |||
AK149865.1 | - | 10 | 1148 | |||
AK151335.1 | - | 10 | 1147 | |||
AK153390.1 | - | 10 | 1147 | |||
AK170688.1 | - | 10 | 1172 | |||
AK171565.1 | - | 10 | 1173 | |||
BC012721.1 | - | 10 | 1210 | 4037046 | ||
BC067198.1 | - | 10 | 1196 | 6826682 | ||
U15635.1 | - | 10 | 1197 |