Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159616.1 | NP_001153088.1 | 3 | 261 | ||
NM_001159617.1 | NP_001153089.1 | 3 | 430 | |||
NM_001159618.1 | NP_001153090.1 | 3 | 340 | |||
NM_001159619.1 | NP_001153091.1 | 2 | 293 | |||
NM_001159620.1 | NP_001153092.1 | 2 | 293 | |||
NM_019543.3 | NP_062416.1 | 3 | 276 | |||
GenBank mRNA | AF216306.1 | - | 3 | 263 | ||
AF216307.1 | - | 3 | 238 | |||
AF216308.1 | - | 2 | 183 | |||
AF216309.1 | - | 3 | 241 | |||
AK004169.1 | - | 3 | 240 | |||
BC038252.1 | - | 3 | 441 | 4913346 | ||
BC061116.1 | - | 3 | 214 | 6593482 | ||
BC061117.1 | - | 2 | 252 | |||
BC096569.1 | - | 3 | 268 | 1429742 | ||
BX511452.1 | - | 3 | 341 | |||
BY041400.1 | - | 3 | 261 |