Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253690.1 | NP_001240619.1 | 5 | 2109 | ||
NM_001253691.1 | NP_001240620.1 | 5 | 2128 | |||
NM_019574.3 | NP_062520.1 | 4 | 2036 | |||
GenBank mRNA | AB029397.1 | - | 4 | 2036 | ||
AK018372.1 | - | 4 | 790 | |||
BC026673.1 | - | 4 | 707 | |||
BC043035.1 | - | 3 | 1862 | 5697155 | ||
BC055947.1 | - | 6 | 840 | |||
CA327687.1 | - | 5 | 717 |