Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204275.1 | NP_001191204.1 | 12, 13 | 2014 | ||
NM_001204276.1 | NP_001191205.1 | 13, 14 | 2062 | |||
NM_001204277.1 | NP_001191206.1 | 12, 13 | 1933 | |||
NM_001204278.1 | NP_001191207.1 | 11, 12 | 1903 | |||
NM_001204279.1 | NP_001191208.1 | 11, 12 | 1831 | |||
NM_001204280.1 | NP_001191209.1 | 11, 12 | 1903 | |||
NM_001204281.1 | NP_001191210.1 | 10, 11 | 1466 | |||
NM_019572.3 | NP_062518.2 | 11, 12 | 1915 | |||
GenBank mRNA | AF207749.1 | - | 11, 12 | 1525 | ||
AK028515.1 | - | 10, 11 | 1439 | |||
AK030863.1 | - | 12, 13 | 1815 | |||
AK036586.1 | - | 13, 14 | 1790 | |||
AK037282.1 | - | 11, 12 | 1567 | |||
AK040406.1 | - | 11, 12 | 1617 | |||
AK044287.1 | - | 12, 13 | 1661 | |||
AK088828.1 | - | 11, 12 | 1599 | |||
AK088945.1 | - | 11, 12 | 1659 | |||
AK131123.1 | - | 11, 12 | 1642 | |||
AK146315.1 | - | 11, 12 | 1704 | |||
AK150041.1 | - | 11, 12 | 1627 | |||
BC057332.1 | - | 11, 12 | 1819 | 6826982 |