Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019805.4 | NP_062779.3 | 2 | 293 | ||
GenBank mRNA | AF076607.2 | - | 2 | 257 | ||
AK030775.1 | - | 2 | 258 | |||
AK049968.1 | - | 2 | 231 | |||
AK137286.1 | - | 2 | 257 | |||
AK150764.1 | - | 2 | 257 | |||
AK150891.1 | - | 2 | 257 | |||
AK153114.1 | - | 2 | 257 | |||
AK159523.1 | - | 2 | 225 | |||
AK164202.1 | - | 2 | 262 | |||
BC006635.1 | - | 2 | 246 | 3482261 | ||
BY128328.1 | - | 2 | 294 |