Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146200.1 | NP_666312.1 | 19 | 2530 | ||
GenBank mRNA | AK088076.1 | - | 19 | 2573 | ||
AK152693.1 | - | 19 | 2548 | |||
AK160078.1 | - | 19 | 2550 | |||
AK168160.1 | - | 19 | 2534 | |||
AK168174.1 | - | 19 | 2534 | |||
AK212354.1 | - | 3 | 291 | |||
BC024855.1 | - | 19 | 2530 | 5359060 | ||
BC025032.1 | - | 19 | 2551 | 4947435 | ||
BC080763.1 | - | 6 | 945 | |||
U67328.1 | - | 8 | 1302 |