Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019743.3 | NP_062717.2 | 4 | 493 | ||
GenBank mRNA | AF101779.1 | - | 4 | 550 | ||
AK033024.1 | - | 4 | 532 | |||
BC053016.1 | - | 4 | 493 | 6406584 | ||
BC080287.1 | - | 4 | 576 | 6410301 | ||
BC138324.1 | - | 4 | 475 | 8861344 | ||
BC138325.1 | - | 4 | 475 | 8861345 |