Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019736.3 | NP_062710.2 | 14 | 1344 | ||
GenBank mRNA | AB028898.1 | - | 14 | 1248 | ||
AJ238893.1 | - | 14 | 1299 | |||
AK002892.1 | - | 14 | 1308 | |||
AK009717.1 | - | 14 | 1307 | |||
AK076513.1 | - | 15 | 1398 | |||
AK088082.1 | - | 14 | 1304 | |||
AK168713.1 | - | 14 | 1346 | |||
AK170876.1 | - | 14 | 1307 | |||
BC021763.1 | - | 14 | 1334 | 4458575 |