Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019790.4 | NP_062764.1 | 3, 4 | 830 | ||
GenBank mRNA | AB017270.1 | - | 3, 4 | 793 | ||
AB041565.1 | - | 3, 4 | 834 | |||
AK029307.1 | - | 3, 4 | 830 | |||
AK030053.1 | - | 8, 9 | 2066 | |||
AK135038.1 | - | 3, 4 | 817 | |||
BC034850.1 | - | 3, 4 | 806 | 1397175 |