Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019771.2 | NP_062745.1 | 2 | 417 | ||
GenBank mRNA | AB025406.1 | - | 2 | 137 | ||
AK078898.1 | - | 2 | 191 | |||
BC131926.1 | - | 2 | 134 | 40130203 | ||
CT010216.1 | - | 1 | 103 |