Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019793.3 | NP_062767.3 | 6 | 809 | ||
GenBank mRNA | AB009021.1 | - | 6 | 591 | ||
AB072450.1 | - | 6 | 619 | |||
AF133427.1 | - | 6 | 603 | |||
AF242591.1 | - | 6 | 802 | |||
AK006798.1 | - | 6 | 685 | |||
AK053325.1 | - | 6 | 660 | |||
AK089494.1 | - | 641 | ||||
AK141951.1 | - | 6 | 667 | |||
AK146130.1 | - | 6 | 797 | |||
AK152175.1 | - | 6 | 694 | |||
AK154919.1 | - | 641 | ||||
AK159324.1 | - | 6 | 780 | |||
AK159418.1 | - | 6 | 753 | |||
AK159621.1 | - | 6 | 764 | |||
AK159914.1 | - | 6 | 742 | |||
AK168231.1 | - | 6 | 797 | |||
AK169068.1 | - | 6 | 780 | |||
AK178223.1 | - | 4 | 289 | |||
AK182665.1 | - | 4 | 287 | |||
AK187762.1 | - | 3 | 276 | |||
AK188385.1 | - | 4 | 287 | |||
AK207599.1 | - | 3 | 287 | |||
AK210479.1 | - | 3 | 256 | |||
AK220124.1 | - | 4 | 287 | |||
BC043072.1 | - | 6 | 787 | 6404165 |