Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019776.2 | NP_062750.2 | 5 | 756 | ||
GenBank mRNA | AB021491.1 | - | 5 | 713 | ||
AK034914.1 | - | 4 | 625 | |||
AK088028.1 | - | 5 | 715 | |||
AK133837.1 | - | 5 | 762 | |||
AK159837.1 | - | 5 | 721 | |||
AK161591.1 | - | 5 | 710 | |||
AK162432.1 | - | 5 | 697 | |||
AK167576.1 | - | 5 | 746 | |||
AK178471.1 | - | 1 | 91 | |||
BC007126.1 | - | 5 | 703 | 3592548 | ||
BM944959.1 | - | 4 | 434 |