Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019749.4 | NP_062723.1 | 3, 4 | 630 | ||
GenBank mRNA | AF161587.1 | - | 3, 4 | 350 | ||
AK002879.1 | - | 3, 4 | 385 | |||
AK011731.1 | - | 3, 4 | 295 | |||
AV029091.2 | - | 3, 4 | 401 | |||
BC002126.1 | - | 3, 4 | 393 | 3497484 | ||
BC024621.1 | - | 3, 4 | 413 | 4459651 | ||
BC029329.1 | - | 3, 4 | 332 | |||
BC030350.1 | - | 3, 4 | 639 | 5102114 |