Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019650.3 | NP_062624.2 | 6 | 593 | ||
GenBank mRNA | AF007550.1 | - | 6 | 541 | ||
AK002556.1 | - | 6 | 583 | |||
AK005697.1 | - | 6 | 571 | |||
AK013052.1 | - | 6 | 587 | |||
AK150004.1 | - | 6 | 591 | |||
AK205139.1 | - | 3 | 274 | |||
BC008525.1 | - | 6 | 589 | 3601627 | ||
BC051253.1 | - | 6 | 559 | 6485874 |