Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163780.1 | NP_001157252.1 | 12 | 2031 | ||
NM_019987.2 | NP_064371.2 | 12 | 1959 | |||
GenBank mRNA | AF225918.1 | - | 12 | 1799 | ||
AK052178.1 | - | 5 | 928 | |||
AK087484.1 | - | 12 | 1883 | |||
AK133537.1 | - | 12 | 1898 | |||
AK154185.1 | - | 12 | 2032 | |||
AK173068.1 | - | 7 | 1188 | |||
BC028863.1 | - | 11 | 1716 | 4224269 |