Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019921.3 | NP_064305.2 | 5, 6 | 1151 | ||
GenBank mRNA | AF021833.1 | - | 2, 3 | 505 | ||
AK049399.1 | - | 5, 6 | 1120 | |||
AK051044.1 | - | 3, 4 | 786 | |||
AK083255.1 | - | 5, 6 | 1120 | |||
BC038483.1 | - | 2, 3 | 209 | |||
BC054105.1 | - | 2, 3 | 454 | 30022728 |