Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145454.2 | NP_001138926.1 | 3 | 257 | ||
NM_005563.3 | NP_005554.1 | 3 | 257 | |||
NM_203399.1 | NP_981944.1 | 3 | 233 | |||
NM_203401.1 | NP_981946.1 | 3 | 445 | |||
GenBank mRNA | AB209282.1 | - | 3 | 161 | ||
AB451319.1 | - | 1 | 81 | |||
AB451448.1 | - | 1 | 81 | |||
AK054594.1 | - | 3 | 174 | |||
AK303692.1 | - | 3 | 173 | |||
AK311801.1 | - | 3 | 174 | |||
AV714035.1 | - | 3 | 256 | |||
BC014353.1 | - | 3 | 226 | 3683516 | ||
BC082228.1 | - | 2 | 155 | 2822803 | ||
BF686314.1 | - | 285 | ||||
BG030679.1 | - | 3 | 228 | |||
BG493669.1 | - | 3 | 257 | |||
BM563894.1 | - | 3 | 278 | |||
BQ428400.1 | - | 3 | 478 | |||
BX647885.1 | - | 3 | 175 | |||
J04991.1 | - | 3 | 184 | |||
X53305.1 | - | 3 | 162 | |||
Z11566.1 | - | 3 | 172 |