Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020042.2 | NP_064426.2 | 11 | 1942 | ||
NM_028464.1 | NP_082740.1 | 11 | 1957 | |||
GenBank mRNA | AK157231.1 | - | 11 | 1898 | ||
BC027444.1 | - | 3 | 868 | |||
BC034066.1 | - | 7 | 1306 | |||
BC049914.1 | - | 9 | 1585 | |||
BC147411.1 | - | 2 | 786 | |||
BC147412.1 | - | 2 | 771 | |||
BC172067.1 | - | 2 | 771 |