Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igdcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290315.1 | NP_001277244.1 | 12 | 2205 | ||
NM_020043.3 | NP_064427.2 | 12 | 2202 | |||
GenBank mRNA | AB052620.1 | - | 12 | 2202 | ||
AB052621.1 | - | 12 | 2205 | |||
AF176694.1 | - | 12 | 2153 | |||
AK122535.1 | - | 2281 | ||||
AK143256.1 | - | 11 | 2664 | |||
BU701299.1 | - | 2 | 195 |