Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145454.2 | NP_001138926.1 | 4 | 495 | ||
NM_005563.3 | NP_005554.1 | 4 | 495 | |||
NM_203399.1 | NP_981944.1 | 4 | 471 | |||
NM_203401.1 | NP_981946.1 | 4 | 683 | |||
GenBank mRNA | AB209282.1 | - | 3 | 2357 | ||
AB451319.1 | - | 2 | 319 | |||
AB451448.1 | - | 2 | 319 | |||
AK054594.1 | - | 5 | 391 | |||
AK303692.1 | - | 4 | 411 | |||
AK311801.1 | - | 4 | 412 | |||
BC014353.1 | - | 4 | 464 | 3683516 | ||
BC082228.1 | - | 3 | 393 | 2822803 | ||
BF686314.1 | - | 523 | ||||
BG030679.1 | - | 4 | 466 | |||
BG493669.1 | - | 4 | 496 | |||
BM563894.1 | - | 4 | 516 | |||
BQ428400.1 | - | 4 | 716 | |||
BX647885.1 | - | 3 | 2371 | |||
J04991.1 | - | 4 | 422 | |||
X53305.1 | - | 4 | 400 | |||
Z11566.1 | - | 4 | 410 |