Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020044.3 | NP_064428.1 | 12 | 706 | ||
NM_022964.4 | NP_075253.2 | 11 | 670 | |||
GenBank mRNA | AF257136.1 | - | 11 | 650 | ||
AK134721.1 | - | 9 | 398 | |||
AK138953.1 | - | 12 | 665 | |||
AK143533.1 | - | 11 | 671 | |||
AK144261.1 | - | 12 | 645 | |||
AK162321.1 | - | 11 | 629 | |||
AY190024.1 | - | 12 | 671 | |||
BC005804.1 | - | 11 | 599 | 3154073 |