Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174047.1 | NP_001167518.1 | 13 | 1487 | ||
NM_001174048.1 | NP_001167519.1 | 13 | 1487 | |||
NM_001174049.1 | NP_001167520.1 | 13 | 1487 | |||
NM_001174050.1 | NP_001167521.1 | 13 | 1487 | |||
NM_020263.3 | NP_064659.2 | 13 | 1487 | |||
GenBank mRNA | AB093246.1 | - | 13 | 1142 | ||
AF247139.1 | - | 14 | 1741 | |||
AF247141.1 | - | 12 | 1082 | |||
AK044603.1 | - | 13 | 1303 | |||
AY502107.1 | - | 13 | 1415 | |||
BC056389.1 | - | 13 | 1487 | 6854935 | ||
BC158058.1 | - | 13 | 1318 | 9007358 |