Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122992.1 | NP_001116464.1 | 7 | 622 | ||
NM_020273.2 | NP_064669.2 | 6 | 559 | |||
GenBank mRNA | AF210433.1 | - | 5 | 520 | ||
AK134349.1 | - | 6 | 560 | |||
BC046983.1 | - | 7 | 593 | 6477130 | ||
BU701415.1 | - | 5 | 406 | |||
BY732360.1 | - | 7 | 597 | |||
DV660282.1 | - | 5 | 477 |