Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053080.3 | NP_444310.3 | 7, 8 | 843 | ||
GenBank mRNA | AF152359.1 | - | 7, 8 | 798 | ||
AF246711.1 | - | 7, 8 | 791 | |||
AF253409.1 | - | 7, 8 | 830 | |||
AF280404.1 | - | 7, 8 | 795 | |||
AK147006.1 | - | 7, 8 | 844 | |||
BC058277.1 | - | 7, 8 | 787 | 6515355 |