Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005564.4 | NP_005555.2 | 5 | 591 | ||
GenBank mRNA | AK301694.1 | - | 5 | 568 | ||
AK316217.1 | - | 5 | 568 | |||
BC033089.1 | - | 5 | 555 | 5421124 | ||
BX644845.1 | - | 5 | 604 | |||
CA454137.1 | - | 5 | 553 | |||
CR542092.1 | - | 5 | 495 | |||
EU644752.1 | - | 5 | 501 | |||
S75256.1 | - | 5 | 435 | |||
X83006.1 | - | 5 | 558 |