Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286630.1 | NP_001273559.1 | 5 | 804 | ||
NM_020508.4 | NP_065254.3 | 5 | 760 | |||
NM_198094.2 | NP_932762.2 | 5 | 750 | |||
GenBank mRNA | AF273217.1 | - | 4 | 675 | ||
AF461395.1 | - | 5 | 736 | |||
AF461396.1 | - | 5 | 736 | |||
AK042431.1 | - | 6 | 978 | |||
AK147545.1 | - | 5 | 769 | |||
AK154514.1 | - | 4 | 735 | |||
AK219391.1 | - | 3 | 276 | |||
BC011541.1 | - | 4 | 596 | |||
BC013666.1 | - | 4 | 733 | |||
BC082782.1 | - | 4 | 548 | |||
BC103773.1 | - | 4 | 674 | |||
BC138834.1 | - | 5 | 730 | 8861856 | ||
BC138835.1 | - | 5 | 729 | 8861857 |