Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018758.2 | NP_061228.1 | 10, 11 | 1903 | ||
GenBank mRNA | AF070975.1 | - | 10, 11 | 1857 | ||
AK045965.1 | - | 10, 11 | 1904 | |||
AK169462.1 | - | 10, 11 | 1887 | |||
AK170182.1 | - | 10, 11 | 1851 | |||
BC005423.1 | - | 10, 11 | 1835 | 3487448 | ||
BC009666.1 | - | 10, 11 | 1847 | 3490710 |