Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB162929.1 | - | 10 | 1086 | ||
AF190699.1 | - | 10 | 1158 | |||
AF190991.1 | - | 10 | 1158 | |||
AK015486.1 | - | 9 | 809 | |||
AK018148.1 | - | 10 | 1150 | |||
AK031550.1 | - | 10 | 1186 | |||
AK042758.1 | - | 10 | 939 | |||
AK043998.1 | - | 10 | 1206 | |||
AK048413.1 | - | 10 | 1157 | |||
AK049903.1 | - | 9 | 892 | |||
AK081981.1 | - | 10 | 1160 | |||
AK083126.1 | - | 9 | 1283 | |||
BC046310.1 | - | 10 | 1144 | 6307420 | ||
BC065694.1 | - | 10 | 1150 | 6813365 |