Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199275.2 | NP_001186204.1 | 11 | 1313 | ||
NM_001199276.2 | NP_001186205.1 | 9 | 983 | |||
NM_001271455.1 | NP_001258384.1 | 10 | 1036 | |||
NM_001271456.1 | NP_001258385.1 | 11 | 1208 | |||
NM_021327.4 | NP_067302.2 | 10 | 1155 | |||
GenBank mRNA | AJ242777.1 | - | 9 | 955 | ||
AJ242778.1 | - | 10 | 1127 | |||
AK135338.1 | - | 10 | 1134 | |||
AK156131.1 | - | 10 | 1009 | |||
AK170841.1 | - | 9 | 961 | |||
AK172890.1 | - | 941 | ||||
AY012159.2 | - | 7 | 725 | |||
BC008186.1 | - | 10 | 1072 | 3589311 | ||
BC008665.1 | - | 11 | 1179 | 3598934 | ||
CB599739.1 | - | 1 | 42 |