Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021028.3 | NP_066356.3 | 8 | 606 | ||
GenBank mRNA | AF105217.1 | - | 8 | 581 | ||
AJ249341.2 | - | 8 | 584 | |||
AK041651.1 | - | 8 | 597 | |||
AK051286.1 | - | 7 | 526 | |||
AK087557.1 | - | 8 | 607 | |||
AK154160.1 | - | 8 | 600 | |||
AK178795.1 | - | 4 | 231 | |||
AK187899.1 | - | 4 | 235 | |||
AK193655.1 | - | 4 | 231 | |||
AK196494.1 | - | 4 | 231 | |||
AK216095.1 | - | 4 | 231 | |||
BC019982.1 | - | 8 | 598 | 4159742 |