Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hacd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021345.2 | NP_067320.2 | 9 | 979 | ||
GenBank mRNA | AK050556.1 | - | 9 | 973 | ||
AK051735.1 | - | 9 | 938 | |||
AK052865.1 | - | 9 | 976 | |||
AK159494.1 | - | 9 | 945 | |||
BC031755.1 | - | 9 | 963 | 4488391 | ||
BC057023.1 | - | 9 | 955 | 6813640 |