Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178922.3 | NP_849253.2 | 3 | 456 | ||
GenBank mRNA | AJ868292.1 | - | 2 | 420 | ||
AK028357.1 | - | 2 | 268 | |||
AK084975.1 | - | 3 | 379 | |||
AK129266.1 | - | 3 | 442 | |||
AK156332.1 | - | 3 | 472 | |||
AK157276.1 | - | 3 | 472 | |||
BC065124.1 | - | 455 | 30537019 | |||
BQ965108.1 | - | 3 | 463 |