Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021394.2 | NP_067369.2 | 6 | 1009 | ||
GenBank mRNA | AF136520.1 | - | 6 | 977 | ||
AK157215.1 | - | 5 | 2350 | |||
AX775969.1 | - | 6 | 963 | |||
AX775971.1 | - | 6 | 963 | |||
BC020033.1 | - | 6 | 979 | 4007636 |