Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trem3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021407.3 | NP_067382.1 | 2 | 488 | ||
GenBank mRNA | AF241220.1 | - | 2 | 440 | ||
AK079407.1 | - | 2 | 413 | |||
BC127035.1 | - | 2 | 423 | 40049608 | ||
BC127036.1 | - | 2 | 423 | 40049609 |