Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021424.2 | NP_067399.2 | 2 | 207 | ||
GenBank mRNA | AF239762.1 | - | 2 | 139 | ||
AF270977.1 | - | 2 | 139 | |||
AF297665.1 | - | 2 | 154 | |||
AK147479.1 | - | 799 | ||||
BC060694.1 | - | 2 | 207 | 6405088 |