Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dexi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021428.4 | NP_067403.2 | 1 | 467 | ||
GenBank mRNA | AF152470.2 | - | 1 | 168 | ||
AK007644.1 | - | 1 | 468 | |||
AK032104.1 | - | 468 | ||||
AK183993.1 | - | 1 | 36 | |||
BC017551.1 | - | 1 | 454 | 4910742 |