Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021429.3 | NP_067404.2 | 3 | 284 | ||
GenBank mRNA | AJ132192.1 | - | 3 | 278 | ||
AK040654.1 | - | 3 | 284 | |||
AK084802.1 | - | 3 | 277 | |||
AK154696.1 | - | 3 | 280 | |||
AK170838.1 | - | 3 | 271 | |||
BC095977.1 | - | 3 | 281 |