Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177772.1 | NP_001171243.1 | 13 | 2176 | ||
NM_021471.2 | NP_067446.1 | 13 | 2176 | |||
GenBank mRNA | AK045782.1 | - | 3517 | |||
AK048926.1 | - | 13 | 2146 | |||
AK161010.1 | - | 3484 | ||||
AY007379.1 | - | 13 | 2012 | |||
BC078456.1 | - | 13 | 2085 | 30360663 | ||
BE865248.1 | - | 4 | 439 |