Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153574.2 | NP_705802.1 | 14 | 1690 | ||
GenBank mRNA | AJ510136.1 | - | 14 | 1676 | ||
AK036291.1 | - | 7 | 897 | |||
AK052421.1 | - | 14 | 1631 | |||
BC005554.1 | - | 14 | 1595 | |||
BC054053.1 | - | 14 | 1664 | |||
BC060978.1 | - | 14 | 1627 | |||
BC080861.1 | - | 14 | 1690 | 30357292 |