Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021518.3 | NP_067493.1 | 5 | 569 | ||
GenBank mRNA | AB232585.1 | - | 5 | 350 | ||
AK042862.1 | - | 5 | 539 | |||
AK079071.1 | - | 5 | 570 | |||
AK135421.1 | - | 5 | 553 | |||
AK146179.1 | - | 5 | 554 | |||
AK150030.1 | - | 5 | 601 | |||
AK159607.1 | - | 5 | 590 | |||
AK160333.1 | - | 5 | 570 | |||
AK163320.1 | - | 5 | 552 | |||
AK181047.1 | - | 1 | 73 | |||
AK208722.1 | - | 1 | 73 | |||
AK212465.1 | - | 1 | 73 | |||
AK212917.1 | - | 1 | 95 | |||
AK216186.1 | - | 1 | 73 | |||
AK219567.1 | - | 1 | 73 | |||
BC058382.1 | - | 5 | 567 | 6820798 | ||
X95403.1 | - | 5 | 557 |